siRNA Details
Gene Symbol: Mir99ahg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033222.1 | - | 799 | |||
AK082961.1 | - | 1299 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB649986.1 | - | 1 | 225 | Chr. 16 - 77594902 - 77773427 [+] on Build GRCm38 |
