siRNA Details
Gene Symbol: Gm15867 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK145301.1 | - | 2 | 238 | ||
BB848649.1 | - | 2 | 234 | |||
BE534397.1 | - | 190 | ||||
BE650007.1 | - | 2 | 133 | |||
BY735478.1 | - | 2 | 236 |
