siRNA Details
Gene Symbol: Gm15867 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK145301.1 | - | 2 | 362 | ||
BB836481.1 | - | 2 | 366 | |||
BE534397.1 | - | 314 | ||||
BE650007.1 | - | 2 | 257 | |||
BY735478.1 | - | 2 | 360 |
