siRNA Details
Gene Symbol: Gm15867 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK145301.1 | - | 1, 2 | 158 | ||
BB836481.1 | - | 1, 2 | 163 | |||
BB848649.1 | - | 1, 2 | 154 | |||
BE534397.1 | - | 110 | ||||
BE650007.1 | - | 1, 2 | 53 | |||
BY735478.1 | - | 1, 2 | 156 |
