siRNA Details
Gene Symbol: Gm29682 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033085.1 | - | 2 | 106 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB624123.1 | - | 2 | 106 | Chr. 8 - 104090207 - 104105873 [-] on Build GRCm38 |
