siRNA Details
Gene Symbol: Gm15446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040366.1 | 1 | 56 | |||
GenBank mRNA | AK142681.1 | - | 1 | 56 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY730939.1 | - | 1 | 103 | Chr. 5 - 109933541 - 109940545 [+] on Build GRCm38 |
