siRNA Details
Gene Symbol: Gm40421 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131776.1 | - | 128 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707213.1 | - | 2 | 123 | Chr. 12 - 50985300 - 51004868 [-] on Build GRCm38 |
