siRNA Details
Gene Symbol: Gm11587 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK135711.1 | - | 2 | 659 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB698797.1 | - | 1 | 39 | Chr. 1 - 61306644 - 61307746 [+] on Build GRCm38 |
