siRNA Details
Gene Symbol: Gm30052 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK135717.1 | - | 2 | 150 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ052506.1 | - | 2 | 149 | Chr. 8 - 113668206 - 113676458 [-] on Build GRCm38 |
