siRNA Details
Gene Symbol: 8430439B09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020238.1 | - | 215 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY720611.1 | - | 1 | 215 | Chr. X - 11493926 - 11494291 [+] on Build GRCm38 |
