siRNA Details
Gene Symbol: 8430411E10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020233.1 | - | 1 | 80 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY720523.1 | - | 1 | 80 | Chr. 14 - 105167766 - 105168076 [-] on Build GRCm38 |
