siRNA Details
Gene Symbol: 8430426J06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_077229.1 | 1 | 154 | |||
GenBank mRNA | AK020235.1 | - | 1 | 71 | ||
BE628871.1 | - | 1 | 104 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY720575.1 | - | 1 | 68 | Chr. 15 - 81242957 - 81247885 [-] on Build GRCm38 |
