siRNA Details
Gene Symbol: 4930461G14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040736.1 | 3 | 669 | |||
GenBank mRNA | AK015493.1 | - | 3 | 669 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715041.1 | - | 3 | 668 | Chr. 9 - 58455173 - 58469618 [+] on Build GRCm38 |
