siRNA Details
Gene Symbol: 1600025M17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038168.1 | 1 | 66 | |||
GenBank mRNA | AA098421.1 | - | 1 | 66 | ||
AK005539.1 | - | 1 | 21 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY705750.1 | - | 1 | 21 | Chr. X - 56317654 - 56320664 [+] on Build GRCm38 |
