siRNA Details
Gene Symbol: 3200001D21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045888.2 | 3 | 1542 | |||
GenBank mRNA | AK014278.1 | - | 3 | 1543 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 3200001D21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045888.2 | 3 | 1542 | |||
GenBank mRNA | AK014278.1 | - | 3 | 1543 |