siRNA Details
Gene Symbol: Nhsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041760.1 | - | 4 | 416 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB637868.1 | - | 4 | 413 | Chr. 10 - 18407640 - 18419525 [+] on Build GRCm38 |
