siRNA Details
Gene Symbol: 9530022J15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020566.1 | - | 1 | 234 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY722037.1 | - | 1 | 232 | Chr. 12 - 12293447 - 12294067 [+] on Build GRCm38 |
