siRNA Details
Gene Symbol: E230015B07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042791.1 | - | 4 | 389 | ||
AK054052.1 | - | 1359 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: E230015B07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042791.1 | - | 4 | 389 | ||
AK054052.1 | - | 1359 |