siRNA Details
Gene Symbol: E230015B07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042791.1 | - | 6 | 823 | ||
AK054052.1 | - | 1725 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_177057.1 | - | 1725 | Chr. - - [-] on Build |
