siRNA Details
Gene Symbol: Stamos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038162.1 | 1 | 84 | |||
GenBank mRNA | AK014986.1 | - | 1 | 85 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CN833366.1 | - | 1 | 112 | Chr. 2 - 14070523 - 14073959 [-] on Build GRCm38 |
