siRNA Details
Gene Symbol: 4930477J04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015580.1 | - | 1 | 299 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715130.1 | - | 1 | 299 | Chr. 18 - 36320906 - 36321492 [-] on Build GRCm38 |
