siRNA Details
Gene Symbol: 4930478M09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015588.1 | - | 1 | 33 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715138.1 | - | 1 | 33 | Chr. 5 - 31451513 - 31452331 [-] on Build GRCm38 |
