siRNA Details
Gene Symbol: 4930567J20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040576.1 | 3 | 535 | |||
GenBank mRNA | AK019789.1 | - | 3 | 535 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930567J20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040576.1 | 3 | 535 | |||
GenBank mRNA | AK019789.1 | - | 3 | 535 |