siRNA Details
Gene Symbol: Gm28590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031624.1 | - | 3 | 290 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB621240.1 | - | 3 | 290 | Chr. 1 - 122205145 - 122250480 [+] on Build GRCm38 |
