siRNA Details
Gene Symbol: 5830440H09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017981.1 | - | 1 | 291 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY717897.1 | - | 1 | 291 | Chr. 16 - 50752340 - 50753207 [+] on Build GRCm38 |
