siRNA Details
Gene Symbol: 4930566F21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016234.1 | - | 2 | 583 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715846.1 | - | 2 | 583 | Chr. 5 - 31487299 - 31495179 [-] on Build GRCm38 |
