siRNA Details
Gene Symbol: 5033421B08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017181.1 | - | 2 | 341 | ||
AK050516.1 | - | 3 | 629 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716911.1 | - | 2 | 341 | Chr. 12 - 8549621 - 8562650 [+] on Build GRCm38 |
