siRNA Details
Gene Symbol: 9530022L04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020567.1 | - | 1 | 13 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY722038.1 | - | 11 | Chr. 3 - 30957316 - 30957636 [-] on Build GRCm38 |
