siRNA Details
Gene Symbol: 2810452K05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013332.1 | - | 1 | 750 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ243472.1 | - | 1 | 405 | Chr. 4 - 129624255 - 129624723 [-] on Build GRCm38 |
