siRNA Details
Gene Symbol: 9530001D17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020532.1 | - | 1 | 215 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721963.1 | - | 1 | 215 | Chr. 13 - 54741952 - 54742336 [+] on Build GRCm38 |
