siRNA Details
Gene Symbol: 9430099M06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020530.1 | - | 1 | 95 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721960.1 | - | 1 | 94 | Chr. 8 - 102706138 - 102706790 [-] on Build GRCm38 |
