siRNA Details
Gene Symbol: 9430099N05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020531.1 | - | 1 | 299 | ||
AK042099.1 | - | 1 | 926 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721961.1 | - | 1 | 297 | Chr. 12 - 110471416 - 110471988 [-] on Build GRCm38 |
