siRNA Details
Gene Symbol: Gm38604 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK138291.1 | - | 420 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CJ057246.1 | - | 3 | 420 | Chr. 13 - 78124579 - 78183159 [-] on Build GRCm38 |
