siRNA Details
Gene Symbol: 1700025N23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040523.1 | 4 | 393 | |||
GenBank mRNA | AK006361.1 | - | 4 | 393 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706428.1 | - | 404 | Chr. - - [-] on Build |
