siRNA Details
Gene Symbol: 1700025N21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006360.1 | - | 1 | 160 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706427.1 | - | 1 | 160 | Chr. 10 - 83139150 - 83142038 [-] on Build GRCm38 |
