siRNA Details
Gene Symbol: Sntg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080155.1 | - | 2 | 1223 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB221905.2 | - | 2 | 183 | Chr. 1 - 8950572 - 9298236 [-] on Build GRCm38 |
