siRNA Details
Gene Symbol: Gm38575 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038448.1 | - | 445 | |||
AK165297.1 | - | 376 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB632197.1 | - | 2 | 445 | Chr. 10 - 85231572 - 85266392 [-] on Build GRCm38 |
