siRNA Details
Gene Symbol: Tenm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020393.1 | - | 1 | 424 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721501.1 | - | 1 | 424 | Chr. 11 - 36671960 - 36672602 [-] on Build GRCm38 |
