siRNA Details
Gene Symbol: 1700015C17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005987.1 | - | 2 | 138 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706115.1 | - | 2 | 138 | Chr. 3 - 153467214 - 153475565 [-] on Build GRCm38 |
