siRNA Details
Gene Symbol: 2310016D03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045491.1 | 3 | 329 | |||
GenBank mRNA | AK009380.1 | - | 3 | 330 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY709136.1 | - | 3 | 330 | Chr. 12 - 30411438 - 30467358 [-] on Build GRCm38 |
