siRNA Details
Gene Symbol: Zfp605 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK051473.1 | - | 3, 4 | 509 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB656693.1 | - | 3, 4 | 506 | Chr. 5 - 110110338 - 110123845 [+] on Build GRCm38 |
