siRNA Details
Gene Symbol: D930048G16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK086736.1 | - | 2 | 1935 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB528815.2 | - | 1 | 512 | Chr. 17 - 23793569 - 23794241 [+] on Build GRCm38 |
