siRNA Details
Gene Symbol: Gm15764 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC046487.1 | - | 3 | 446 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE914768.1 | - | 3 | 426 | Chr. 16 - 18416702 - 18421983 [+] on Build GRCm38 |
