siRNA Details
Gene Symbol: 3110013M02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014048.1 | - | 1 | 200 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY713044.1 | - | 1 | 198 | Chr. 17 - 85726686 - 85727018 [+] on Build GRCm38 |
