siRNA Details
Gene Symbol: Gm16758 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC066032.1 | - | 471 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CB246473.1 | - | 1 | 471 | Chr. 17 - 29890084 - 29891827 [+] on Build GRCm38 |
