siRNA Details
Gene Symbol: 2310031A07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009549.1 | - | 2 | 733 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY709238.1 | - | 2 | 733 | Chr. 11 - 46440450 - 46447265 [-] on Build GRCm38 |
