siRNA Details
Gene Symbol: AU015791 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102381.1 | 4 | 405 | |||
GenBank mRNA | AK139027.1 | - | 1 | 910 | ||
BG083309.2 | - | 4 | 405 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV381755.2 | - | 2 | 123 | Chr. 12 - 105513734 - 105515246 [+] on Build GRCm38 |
