siRNA Details
Gene Symbol: Khnyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076856.1 | - | 1 | 1997 | ||
AK122247.1 | - | 8 | 4513 | |||
BC034330.1 | - | 1 | 1480 |
Gene Symbol: Sdr39u1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI843700.1 | - | 1 | 138 |
