siRNA Details
Gene Symbol: Gm31077 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK144897.1 | - | 4 | 506 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY734578.1 | - | 4 | 506 | Chr. 16 - 23520319 - 23543821 [+] on Build GRCm38 |
