siRNA Details
Gene Symbol: Gm41764 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK078684.1 | - | 294 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB623986.1 | - | 4 | 294 | Chr. 18 - 68101179 - 68118376 [-] on Build GRCm38 |
